Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCR9 Recombinant Rat Monoclonal Antibody (9B1), Invitrogen™

Rat Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA548298

Encompass

Catalog No. PIMA548298


Add to Cart

Description

Description

Specificity: 9B1 antibody recognizes CCR9 at the N-terminus. CCR9 is a member of the G protein coupled receptor family and is involved in T cell development in the thymus and in gut-associated immune response. Mouse CCR9 is highly expressed on different stages of thymocytes and upregulated on CD4+CD8+ cells.

CCR9 Recombinant Monoclonal antibody specifically detects CCR9 in Human samples. It is validated for Flow Cytometry, Neutralization
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CCR9
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P51681
Rat
Affinity chromatography
RUO
1234
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG2a
Flow Cytometry, Neutralization
9B1
Unconjugated
CCR5
A130091K22Rik; C-C chemokine receptor type 9; C-C CKR-9; C-C motif chemokine receptor 9; CC-CKR-9; Ccr9; CCR-9; CDw199; CDw199 antigen; chemokine (C-C motif) receptor 9; chemokine C-C receptor 10; Cmkbr10; G protein-coupled receptor 28; GPR28; GPR-9-6; G-protein coupled receptor 28
This antibody was raised by immunizing mice with CCR5.
200 μg
Primary
Human
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit