Learn More
CCRL1 Mouse anti-Human, BV421, Clone: 13E11, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566601
Catalog No. BDB566601
Add to cart
Description
Description
CCRL1 Monoclonal antibody specifically detects CCRL1 in Human samples. It is validated for Flow Cytometry,Immunofluorescence.
Specifications
Specifications
CCRL1
13E11
Brilliant Violet 421
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry, Immunofluorescence
0.2 mg/mL
Purified
IgG2b, κ
RUO
Primary
Human