Learn More
CCRL2/CRAM-A/B Mouse anti-Human, Alexa Fluor™ 594, Clone: MM0158-5A15, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP212137AF594
Catalog No. NP212137A94
Add to cart
Description
Description
CCRL2/CRAM-A/B Monoclonal antibody specifically detects CCRL2/CRAM-A/B in Human samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Paraffin).
Specifications
Specifications
CCRL2/CRAM-A/B
MM0158-5A15
Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin
C-C chemokine receptor-like 2, CCR11, CCR6, chemokine (C-C motif) receptor-like 2, Chemokine receptor CCR11, Chemokine receptor X, CKRXMGC116710, CRAM, CRAM-A, CRAM-B, FLJ55815, HCRMGC34104, Putative MCP-1 chemokine receptor
Mouse
IgG1
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
CCRL2
Human CCRL2
0.1 mL
Primary
9034