Learn More
CCRN4L Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB29991
Catalog No. 89-033-279
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human CCRN4L.
  • High specificity
  • High reactivity
Specifications
Specifications
CCRN4L
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user.
NM_012118
CCR4L|MGC142054|MGC142060|MGC4120817|MGC78549|NOC
Rabbit
Protein A purification
RUO
Primary
25819
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human CCRN4L.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
CCRN4L
A synthetic peptide corresponding to N-terminus of human CCRN4L.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)