Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCS Mouse anti-Human, Clone: 3A1, Abnova™

Mouse monoclonal antibody raised against recombinant human CCS.

Manufacturer:  Abnova Corporation MAB12984

Catalog No. 89-933-736


Add to cart

Description

Description

Copper chaperone for superoxide dismutase specifically delivers Cu to copper/zinc superoxide dismutase and may activate copper/zinc superoxide dismutase through direct insertion of the Cu cofactor. [provided by RefSeq]

Specifications

Specifications

CCS
3A1
Mouse monoclonal antibody raised against recombinant human CCS.
In HEPES, 0.15M NaCl (50% glycerol, 0.01% BSA, 0.03% sodium azide).
MGC138260
Mouse
IgG2a, kappa
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Immunoprecipitation, Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:200) Immunoprecipitation Western Blot (1:2000) The optimal working dilution should be determined by the end user.
Liquid
CCS
Recombinant protein corresponding to human CCS.
Ammonium sulfate precipitation
RUO
Primary
9973
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit