Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT 031374 hydrobromide, Tocris Bioscience™

Click to view available options
Quantity:
10mg
50mg
This item is not returnable. View return policy
1219184-91-4
C23H20BrN3O
434.337
MPILENOYNNNGKO-UHFFFAOYSA-N
cct 031374 hydrobromide,cct031374 hydrobromide hplc,1-1,1'-biphenyl-4-yl-2-2,3-dihydro-9h-imidazo 1,2-a benzimidazol-9-yl-ethanone hydrobromide,1-1,1'-biphenyl-4-yl-2-2,3-dihydro-9h-imidazo 1,2-a benzimidazol-9-yl ethanone hydrobromide,1-1,1'-biphenyl-4-yl-2-2,5,7-triazatricyclo 6.4.0.0 2 ,? dodeca-1 12 ,5,8,10-tetraen-7-yl ethanone hydrobromide,1-1,1'-biphenyl-4-yl-2-2,5,7-triazatricyclo 6.4.0.0_,? dodeca-1 12 ,5,8,10-tetraen-7-yl ethanone hydrobromide
18554738
2-(1,2-dihydroimidazo[1,2-a]benzimidazol-4-yl)-1-(4-phenylphenyl)ethanone;hydrobromide
C1CN2C3=CC=CC=C3N(C2=N1)CC(=O)C4=CC=C(C=C4)C5=CC=CC=C5.Br
This item is not returnable. View return policy
1219184-91-4
C23H20BrN3O
434.337
MPILENOYNNNGKO-UHFFFAOYSA-N
cct 031374 hydrobromide,cct031374 hydrobromide hplc,1-1,1'-biphenyl-4-yl-2-2,3-dihydro-9h-imidazo 1,2-a benzimidazol-9-yl-ethanone hydrobromide,1-1,1'-biphenyl-4-yl-2-2,3-dihydro-9h-imidazo 1,2-a benzimidazol-9-yl ethanone hydrobromide,1-1,1'-biphenyl-4-yl-2-2,5,7-triazatricyclo 6.4.0.0 2 ,? dodeca-1 12 ,5,8,10-tetraen-7-yl ethanone hydrobromide,1-1,1'-biphenyl-4-yl-2-2,5,7-triazatricyclo 6.4.0.0_,? dodeca-1 12 ,5,8,10-tetraen-7-yl ethanone hydrobromide
18554738
2-(1,2-dihydroimidazo[1,2-a]benzimidazol-4-yl)-1-(4-phenylphenyl)ethanone;hydrobromide
C1CN2C3=CC=CC=C3N(C2=N1)CC(=O)C4=CC=C(C=C4)C5=CC=CC=C5.Br

Inhibits TCF-dependent transcription; lowers β-catenin levels

Inhibitor of TCF-dependent transcription. Blocks BIO -induced β-catenin stabilization; reduces nuclear and cytosolic β-catenin levels in mouse L-cells. Inhibits growth and reduces TCF-dependent transcription in SW480 colon carcinoma cells.
Quantity 50mg
Formula Weight 434.33
Percent Purity >98%
Chemical Name or Material CCT 031374 hydrobromide

Recommended Storage : Desiccate at Room Temperature