Learn More
CCT2 Rat anti-Human, Mouse, Clone: PK/8/4/4i/2f, Abnova™
Rat monoclonal antibody raised against mouse Cct2.
Manufacturer:  Abnova Corporation MAB12595
Catalog No. 89-933-737
Add to cart
Description
Description
Specifications
Specifications
CCT2
PK/8/4/4i/2f
Rat monoclonal antibody raised against mouse Cct2.
In PBS, 10mM Na3PO4, 50mM NaCl, pH 7.2 (0.1mM PMSF)
Cctb
Rat
IgG2b
50 ug
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human, Murine
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
Cct2
Mouse Cct2 peptide-PPD conjugated.
Protein G purification
RUO
Primary
12461