Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CCT5 Mouse anti-Human, Hamster, DyLight 405, Clone: 639, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243680V

Catalog No. NBP243680V


Add to cart

Description

Description

CCT5 Monoclonal antibody specifically detects CCT5 in Human, Hamster samples. It is validated for Western Blot.
Specifications

Specifications

CCT5
639
Western Blot
Purified
CCT5
Recombinant protein encompassing a sequence within the center region of human TCP1 epsilon. The exact sequence is proprietary.
Protein G purified
Core ESC Like Genes, Stem Cell Markers
Primary
22948
Western Blot
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
chaperonin containing TCP1, subunit 5 (epsilon), epsilon subunit
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Hamster
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit