Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CCT5 Rabbit, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | CCT5 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP156769
|
Novus Biologicals
NBP156769 |
100 μL |
Each of 1 for $436.00
|
|
Description
CCT5 Polyclonal specifically detects CCT5 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
CCT5 | |
Unconjugated | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
chaperonin containing TCP1, subunit 5 (epsilon), epsilon subunit | |
CCT5 | |
IgG | |
Affinity Purified | |
This product is specific to Subunit or Isoform: epsilon. |
Polyclonal | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
P48643 | |
22948 | |
Synthetic peptides corresponding to CCT5(chaperonin containing TCP1, subunit 5 (epsilon)) The peptide sequence was selected from the N terminal of CCT5. Peptide sequence NDGATILSMMDVDHQIAKLMVELSKSQDDEIGDGTTGVVVLAGALLEEAE. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title