Learn More
Aviva Systems Biology Corp CCT6B Antibody - N-terminal region (ARP53660_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53660_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCT6B. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CCT6B. Alternate names: CCT-zeta-2, CCTZ-2, Cctz2, TCP-1-zeta-2, TSA303ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-61082