Learn More
Aviva Systems Biology Corp CCT7 Antibody - middle region (ARP51808_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP51808_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CCT7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human CCT7. Alternate names: CCT-ETA, Ccth, MGC110985, Nip7-1, TCP-1-eta, CCTH, CCTETA, NIP7-1, TCP1ETAApplicationWB, IHC. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-60621