Learn More
Abnova™ CCT8 293T Cell Transient Overexpression Lysate (Denatured) (T04)
Human overexpression lysate
Manufacturer:  Abnova™ H00010694T04
Catalog No. 89-938-576
Description
Description
Specifications
Specifications
Western Blot
CCT8 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 59.6kDa
Transient overexpression cell lysate was tested with anti-CCT8 antibody (Mfr. No. H00010694-B02P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
293T
NM_006585
pCMV-CCT8 full-length
100 μL
Human