Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody.
Supplier: Epredia™ MS728S1
Description
CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.Host Species: Mouse
Clone: 56C6
Isotype: IgG1
Species Reactivity: Human and Rat. Others not tested.
Epitope: Extracellular
Immunogen: Recombinant external domain of CD10 protein
Molecular Weight: 100kDa
Positive Control: Small intestine
Cellular Localization: Membrane
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD10 | |
Monoclonal | |
Unconjugated | |
Recombinant external domain of CD10 protein | |
IVD | |
Primary | |
Concentrated |
Immunohistochemistry (Paraffin) | |
56C6 | |
Mouse | |
500 μL | |
Oncogenesis | |
Human, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction