Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD10, Mouse anti-Human, RD1, Clone: J5, Beckman Coulter™

Mouse Monoclonal Antibody

$397.80 - $685.44

Specifications

Clone J5
Host Species Murine
Conjugate RD1
Isotype IgG2a
Antigen CD10
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
CO6603475 View Documents Coulter Corporation
6603475
100 Tests Each for $685.44
Add to cart
 
CO6604120 View Documents Coulter Corporation
6604120
50 Tests Each for $397.80
Add to cart
 
Description

Description

The CD10 antigen is referred to as the Common Acute Lymphoblastic Leukemia Antigen (CALLA). It is a type II integral membrane protein of 100 kDa, identified as the human membrane-associated neutral endopeptidase (EC3.4.24.11). It is expressed on uncommitted lymphoid precursors. CD10 expression is lost as cells enter the T lineage. In the B lineage, CD10 expression is lost later in ontogeny, as cells acquire surface Ig expression. It is also expressed on activated and proliferating B cells in the germinal center, and on neutrophils as well as on bone marrow stromal cells. It is also expressed on a number of other cells of epithelial origin.
Specifications

Specifications

J5
RD1
CD10
Affinity Purified
Murine
IgG2a
Monoclonal
Primary
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit