Learn More
CD100 (Sema4D) Mouse, anti-Human, R718, Clone: A8, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752138
Catalog No. BDB752138
Add to cart
Description
Description
The A8 monoclonal antibody specifically binds to CD100 which is also known as Semaphorin-4D (SEMA4D). CD100 is a 150 kDa type I transmembrane glycoprotein that belongs to the semaphorin family. CD100 is widely expressed in the nervous and immune systems. Although it is expressed on T cells, its expression is significantly upregulated on activated T cells. It is also expressed on NK cells, B cells, and monocytes. CD100 is an adhesion molecule that functions as a signaling molecule which plays a role in lymphocyte activation. CD100 interaction with CD72 enhances B cell signaling and ultimately promotes B cell activation and survival.
Specifications
Specifications
CD100 (Sema4D)
A8
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Human