Learn More
CD100 (Sema4D) Rat anti-Mouse, PE, Clone: BMA-12, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 565556
Catalog No. BDB565556
Add to cart
Description
Description
CD100 (Sema4D) Monoclonal antibody specifically detects CD100 (Sema4D) in Mouse samples. It is validated for Flow cytometry.
Specifications
Specifications
CD100 (Sema4D)
BMA-12
PE
Purified
Rat
Rat IgG2a
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with R-PE under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Semaphorin-4D; SEM4D; Semaphorin-C-like 2; Semacl2; Semaj; M-Sema G; coll-4
Mouse CD100 Recombinant Protein
50μg
Cell Biology
Primary
Murine
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.