Learn More
CD102 Mouse anti-Human, BB515, Clone: CBR-IC2/2, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565932
Catalog No. BDB565932
Add to cart
Description
Description
CD102 Monoclonal antibody specifically detects CD102 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD102
CBR-IC2/2
Vol. Per test: 5μL
Purified
Mouse
Mouse BALB/c IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Monoclonal
Flow Cytometry
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
ICAM2; ICAM-2; Intercellular adhesion molecule 2
Human ICAM-2 Transfected Cell Line
25 Tests
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.