Learn More
CD102 Rat anti-Mouse, BV480, Clone: 3C4(mIC2/4), BD OptiBuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 746590
Catalog No. BDB746590
Add to cart
Description
Description
CD102 Monoclonal antibody specifically detects CD102 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD102
3C4(mIC2/4)
Brilliant Violet 480
Rat
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2a, κ
RUO
Primary
Mouse