Learn More
CD103 Mouse anti-Human, BV421, Clone: Ber-ACT8, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566257
Catalog No. BDB566257
Add to cart
Description
Description
CD103 Monoclonal antibody specifically detects CD103 in Human samples. It is validated for Flow cytometry.
Specifications
Specifications
CD103
Ber-ACT8
Vol. Per test: 5μL
Purified
Mouse
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions, and unconjugated antibody and free BD Horizon™ BV421 were removed.
Monoclonal
Flow Cytometry
Brilliant Violet 421
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
ITGAE; Integrin alpha-E; ITAE; HML-1 antigen; Mucosal lymphocyte 1; HUMINAE
Human MAPS16 T Cell Line
25 Tests
Cell Biology
Primary
Human
SDS
Documents
Product Certifications
Promotions
Promotions
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.