Learn More
CD103 Mouse anti-Human, PE, Clone: LF61, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD10304
Catalog No. 50-112-1328
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
The CD103 antigen is a 160-130-105 kDa molecule which is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. This human intra epithelial lymphocyte marker is the E integrin chain associated with the b7 subunit. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs.
Specifications
Specifications
CD103
Monoclonal
PE
ITGAE
Liquid
ITGAE
Human CD103
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
LF61
PBS with 10% sucrose, BSA and 0.1% sodium azide
P38570
CD103, HUMINAE,HML-1 antigen; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; integrin alpha-E; integrin alpha-IEL; mucosal lymphocyte 1 antigen
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
3682