Learn More
CD103 Mouse anti-Human, TRI-COLOR®, Clone: LF61, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD10306
Catalog No. 50-113-7500
Add to cart
Description
Description
The CD103 antigen is a 160-130-105 kDa molecule which is expressed on mucosa-associated T lymphocytes and activated cells and on subet of TGF beta -1 cells. This human intra epithelial lymphocyte marker is the E integrin chain associated with the b7 subunit. CD103 antibody stains a few bone marrow cells and, rarely, peripheral blood lymphocytes. CD103 is expressed by 0.5 to 2% of resting lymphocytes in peripheral blood and lymphoid organs.
Specifications
Specifications
CD103, CD103
Monoclonal
TRI-COLOR®
ITGAE
Liquid
ITGAE
Human CD103
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
LF61
PBS with sucrose and 0.1% sodium azide
P38570
CD103, HUMINAE,HML-1 antigen; antigen CD103, human mucosal lymphocyte antigen 1; alpha polypeptide; integrin alpha-E; integrin alpha-IEL; mucosal lymphocyte 1 antigen
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
3682