Learn More
CD103 Mouse anti-Rat, Alexa Fluor 647, Clone: OX-62, BD Pharmingen™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 565286
Catalog No. BDB565286
Add to cart
Description
Description
CD103 Monoclonal antibody specifically detects CD103 in Rat samples. It is validated for Flow cytometry.
Specifications
Specifications
CD103
OX-62
Alexa Fluor 647
Purified
Murine
Mouse BALB/c IgG1, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to Alexa Fluor™ 647 under optimum conditions
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Integrin alpha E; Itgae; Integrin αE; CD103
Density gradient-enriched PVG rat thoracic-duct dendritic cells
50μg
Cell Biology
Primary
Rat
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Reseach Use Only.