Learn More
CD104 (Integrin beta 4) Mouse anti-Human, Clone: 450-9D, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA528118
Catalog No. PIMA528118
Add to cart
Description
Description
This antibody recognizes an extracellular epitope on the CD104 molecule.
Generalized atrophic benign epidermolysis bullosa (GABEB). A non-lethal, adult form of junctional epidermolysis bullosa characterized by life-long blistering of the skin, associated with hair and tooth abnormalities.
Specifications
Specifications
CD104 (Integrin beta 4)
Monoclonal
1 mg/mL
physiological saline with no preservative
P16144
ITGB4
Purified alpha6 beta4 integrin from A431 cells
Protein G
RUO
Antibody
Monoclonal
Human
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation
450-9D
Unconjugated
ITGB4
Liquid
Mouse
IgG1
1 mg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3691