Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ Lab Vision™ CD105/Endoglin/TGF-β 1/3 Receptor Ab-3, Mouse Monoclonal Antibody

Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications.

Specifications

Antigen CD105/Endoglin/TGF-beta1/3 Receptor Ab-3
Clone SN6h
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Conjugate Unconjugated
View More Specs

Products 3
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Formulation Quantity Price Quantity & Availability  
MS1290R1
View Documents
Thermo Scientific™
MS1290R1
Ready-to-use for IHC (UVLP) 1mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1290P0
View Documents
Thermo Scientific™
MS1290P0
Purified Ab with BSA and Azide 100μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
MS1290P1
View Documents
Thermo Scientific™
MS1290P1
Purified Ab with BSA and Azide 500μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in immunohistochemistry, western blotting and flow cytometry applications with Thermo Scientific™ CD105/Endoglin/TGF-β1/3 Receptor Ab-3, Mouse Monoclonal Antibody.

CD105 or Endoglin or GP160 is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists as an O- and N-glycosylated homodimer. Up-regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation-associated endothelial cell marker. CD105 binds to TGF-Beta1 and Beta3 with high affinity but not TGF-Beta2.

Host Species: Mouse

Clone: SN6h

Isotype: IgG1/κ

Species Reactivity: Human. Others not tested.

Epitope: Not determined

Immunogen: Purified GP160 from cell membrane glycoproteins of fresh non-T/non-B acute lymphoblastic leukemia cells.

Molecular Weight: 95 kDa (monomer) and 190kDa (dimer)

Positive Control: HUVEC cells, Tonsil, lymph node and thymus

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
  • Western Blotting
  • Flow Cytometry

Note:

USA: RUO; Int'l: RUO

Specifications

Specifications

CD105/Endoglin/TGF-beta1/3 Receptor Ab-3
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
RUO
Primary
IgG2a kappa
SN6h
Monoclonal
Murine
Angiogenesis
Purified GP160 from cell membrane glycoproteins of fresh non-T/non-B acute lymphoblastic leukemia cells
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit