Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Epredia™ Lab Vision™ CD105/Endoglin/TGFβ 1/3 Receptor Rabbit Polyclonal Antibody with BSA and azide
Obtain accurate and reliable results in immunohistochemistry experiments with Epredia™ CD105/Endoglin/TGF β 1/3 Receptor, Rabbit Polyclonal Antibody.
Supplier: Epredia™ RB9291P
Description
CD105 or Endoglin or GP160 is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists as an O- and N-glycosylated homodimer. Up-regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation-associated endothelial cell marker. CD105 binds to TGF-β1 and β3 with high affinity but not TGF-β2.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human. Others not tested
Epitope: Not known
Immunogen: Recombinant protein encoding aa 26-176 of human CD105
Molecular Weight: 95 kDa (monomer) and 190 kDa (dimer)
Positive Control: Tonsil, Placenta
Cellular Localization: Membrane and cytoplasm
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Specifications
CD105/Endoglin/TGF beta 1/3 Receptor | |
Polyclonal | |
200µg/mL | |
Rabbit | |
1 mL | |
Angiogenesis | |
Human | |
IgG |
Immunohistochemistry (Paraffin) | |
Polyclonal | |
Unconjugated | |
Recombinant protein encoding aa 26-176 of human CD105 | |
IVD | |
Primary | |
Concentrated |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction