Learn More
CD105 Mouse anti-Human, Clone: SN6, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD10500
Catalog No. 50-113-7501
Add to cart
Description
Description
R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinctio
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.
Specifications
Specifications
CD105, CD105
Monoclonal
Unconjugated
ENG
Liquid
ENG
Human CD105 (Endoglin)
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
SN6
PBS with 0.1% sodium azide
P17813
CD105, END, HHT1, ORW, ORW1, CD105 antigen, ENG
Mouse
IgG1
0.5 mL
4° C, store in dark
Primary
2022