Learn More
CD107b Mouse anti-Human, Unconjugated, Clone: H4B4, Proteintech
Mouse Monoclonal Antibody
Supplier: Proteintech Group Inc 650531IG100UG
This item is not returnable.
View return policy
Description
The CD107b antibody from Proteintech is a mouse monoclonal antibody generated with human adherent peripheral blood cells of human CD107b. This antibody recognizes human antigen. The CD107b antibody has been validated for the following applications: IF, FC analysis.
Specifications
CD107b | |
Monoclonal | |
0.1 mg/mL | |
Mouse | |
Primary | |
Human |
Immunofluorescence | |
H4B4 | |
Unconjugated | |
100 μg | |
3920 | |
IgG1 |