Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD109 Mouse anti-Human, Unconjugated, Clone: KU42.33C, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen CD109
Applications ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABS19281MI
View Documents
MilliporeSigma
MABS1928100UG
100 μg
Each for $372.00
Add to cart
 
MABS19282MI
View Documents
MilliporeSigma
MABS192825UG
25 μg
Each for $147.00
Add to cart
 
Description

Description

CD109 Monoclonal antibody specifically detects CD109 in Human samples. It is validated for ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

CD109
Unconjugated
Mouse
CD109 antigen;150 kDa TGF-beta-1-binding protein;C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7;Platelet-specific Gov antigen;p180;r150
BxPC-3 human pancreatic cancer cells.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Purified
Human
CD109, CPAMD7
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit