Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD116 Recombinant Rabbit Monoclonal Antibody (009), APC, Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA546689

Encompass

Catalog No. PIMA546689


Add to Cart

Description

Description

Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CSF2RA (CD116) (Met1-Gly320) and conjugated with APC under optimum conditions, the unreacted APC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze! Sodium azide is toxic to cells and should be disposed of properly. Flush with large volumes of water during disposal.

CD116 Recombinant Monoclonal antibody specifically detects CD116 in Human samples. It is validated for Flow Cytometry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CD116
Recombinant Monoclonal
100 μg/mL
PBS with 0.5% BSA and 0.09% sodium azide
P15509
CSF2RA
Recombinant protein ecoding Human CD116 (Met1-Gly320).
100 Tests
Primary
Human
Antibody
IgG
Flow Cytometry
009
APC
CSF2RA
CD116; CD116 antigen; CDw116; colony stimulating factor 2 receptor alpha chain; colony stimulating factor 2 receptor alpha subunit; colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage); CSF 2R; CSF2R; Csf2ra; CSF2RAX; CSF2RAY; CSF2RX; CSF2RY; Csfgmra; GM CSF R alpha; GM-CSF receptor alpha subunit; GMCSFR; GM-CSFR; GM-CSF-Ra; GM-CSFRalpha; GM-CSF-R-alpha; GMCSFR-alpha; GMR; GMR-alpha; granulocyte-macrophage colony stimulating receptor alpha; granulocyte-macrophage colony-stimulating factor receptor alpha chain; granulocyte-macrophage colony-stimulating factor receptor subunit alpha; MGC3848; MGC4838; SMDP4
Rabbit
Protein A
RUO
1438
4° C, store in dark, DO NOT FREEZE!
Liquid
Recommendations

Recommendations

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit