Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD117 (c-Kit) Mouse anti-Human, BUV395, Clone: YB5.B8, BD OptiBuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 745740
Description
CD117 (c-Kit) Monoclonal antibody specifically detects CD117 (c-Kit) in Human samples. It is validated for Flow Cytometry.Specifications
CD117 (c-Kit) | |
YB5.B8 | |
Brilliant Ultraviolet 395 | |
Mouse | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG1, κ | |
RUO | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title