Learn More
CD117 Mouse anti-Human, BB700, Clone: YB5.B8, BD Horizon™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 566548
 View more versions of this product
Catalog No. BDB566548
Add to cart
Description
Description
CD117 Monoclonal antibody specifically detects CD117 in Human samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD117
YB5.B8
Purified
IgG1, κ
RUO
Primary
Human
Flow Cytometry
BB700
Mouse
100 Tests
Refrigerate (2 to 8°C)
Monoclonal