Learn More
CD117 Rat anti-Mouse, BB700, Clone: 2B8, BD Horizon™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 566414
 View more versions of this product
Catalog No. BDB566414
Add to cart
Description
Description
CD117 Monoclonal antibody specifically detects CD117 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD117
2B8
BB700
Rat
0.1 mg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2b, κ
RUO
Primary
Mouse