Learn More
CD119 (IFN gamma Receptor 1) Mouse anti-Human, FITC, Clone: BB1E2, Invitrogen™
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MA528109
Catalog No. PIMA528109
Add to cart
Description
Description
This antibody recognizes a single protein band of approximately 43 kDa in western blotting under non-reducing conditions against protein comprising the glycosylated extracellular domain of the human interferon-gamma receptor fused to c-myc, and a MW of 27kDa against a recombit protein corresponding to the non-glycosylated domain fused to c-myc.
IFNGR1 may be involved in Immunodeficiency 27B (IMD27B), a form of Mendelian susceptibility to mycobacterial disease, a rare condition caused by impairment of interferon-gamma mediated immunity. It is characterized by predisposition to illness caused by moderately virulent mycobacterial species, such as Bacillus Calmette-Guerin (BCG) vaccine, environmental non-tuberculous mycobacteria, and by the more virulent Mycobacterium tuberculosis. Other microorganisms rarely cause severe clinical disease in individuals with susceptibility to mycobacterial infections, with the exception of Salmonella which infects less than 50% of these individuals. Clinical outcome severity depends on the degree of impairment of interferon-gamma mediated immunity. Some patients die of overwhelming mycobacterial disease with lepromatous-like lesions in early childhood, whereas others develop, later in life, disseminated but curable infections with tuberculoid granulomas. IMD27B commonly presents with recurrent, moderately severe infections with environmental mycobacteria or BCG. Salmonellosis is present in about 5% of patients. The disease is caused by mutations affecting the gene represented in this entry.
Specifications
Specifications
CD119 (IFN gamma Receptor 1)
Monoclonal
0.1 mg/mL
PBS with 1% BSA and 0.09% sodium azide
P15260
IFNGR1
Fusion protein of the extracellular part of the human interferon-gamma receptor (AA: Ala17-Gly245) conjugated to 11 AA of the c-myc proto-oncogene (for affinity purification)
Ion-Exchange Chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
BB1E2
FITC
IFNGR1
Liquid
Mouse
IgG2a
100 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Primary
3459