Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11a/CD18 Rabbit anti-Human, Clone: hu1124 (Efalizumab), Novus Biologicals™
Rabbit Monoclonal Antibody
$229.00 - $516.00
Specifications
Antigen | CD11a/CD18 |
---|---|
Clone | hu1124 (Efalizumab) |
Dilution | Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready |
Applications | Flow Cytometry, Blocking Assay, CyTOF |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP25268625
|
Novus Biologicals
NBP2526860.025MG |
0.025 mg |
Each for $229.00
|
|
NBP25268602
|
Novus Biologicals
NBP2526860.2MG |
0.2 mg |
Each for $516.00
|
|
Description
CD11a/CD18 Monoclonal specifically detects CD11a/CD18 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF-ready, Block/Neutralize.Specifications
CD11a/CD18 | |
Flow Cytometry 1:10 - 1:1000, Blocking/Neutralizing 50 - 200 molar excess, CyTOF-ready | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD11 antigen-like family member A, CD11a antigen, CD11Aantigen CD11A (p180), lymphocyte function-associated antigen 1, alphapolypeptide, integrin alpha-L, integrin gene promoter, integrin, alpha L (antigen CD11A (p180), lymphocyte function-associated antigen1; alpha polypeptide), Leukocyte adhesion glycoprotein LFA-1 alpha chain, Leukocyte function-associated molecule 1 alpha chain, LFA-1, LFA-1 alpha, LFA1A, LFA-1A, lymphocyte function-associated antigen 1 | |
ITGAL | |
IgG | |
Protein A or G purified | |
Binds to human LFA-1. |
hu1124 (Efalizumab) | |
Flow Cytometry, Blocking Assay, CyTOF | |
Unconjugated | |
Rabbit | |
Cytokine Research, Immunology, Signal Transduction | |
PBS with 0.02% Proclin 300. with 0.02% Proclin 300 | |
3683 | |
Efalizumab binds to CD11a, a subunit of leukocyte function antigen-1 (LFA-1), which is expressed on all leukocytes. | |
Primary | |
Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title