Learn More
CD11a Mouse anti-Rat, BB700, Clone: WT.1, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 745795
Catalog No. BDB745795
Add to cart
Description
Description
CD11a Monoclonal antibody specifically detects CD11a in Rat samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD11a
WT.1
BB700
Mouse
50 μg
Refrigerate (2 to 8°C)
Monoclonal
Flow Cytometry
0.2 mg/mL
Purified
IgG2a, κ
RUO
Primary
Rat