Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD11b Mouse anti-Human, Porcine, Rhesus Ma, Cynomolgus, Chimpanzee, Baboon, FITC, Clone: ICRF44, Southern Biotech™

Mouse Monoclonal Antibody
Supplier: Southern Biotechnology Associates 954602
Description
SouthernBiotech anti-human primary antibodies are available in a variety of conjugated formats including Alexa Fluor and Pacific Blue.
Specifications
CD11b | |
Monoclonal | |
Lot specific | |
Supplied in PBS/NaN3 | |
Rheumatoid synovial cells and human monocytes | |
RUO | |
Baboon, Chimpanzee, Cynomolgus, Human, Porcine, Rhesus Monkey | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
ICRF44 | |
FITC | |
Mouse | |
100 Tests | |
Primary | |
Store at 2-8°C; Protect from light |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title