Learn More
CD11b, Mouse anti-Human, Alexa Fluor 350, Clone: ITGAM/3340, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP234573AF350
Catalog No. NB006172
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD11b Monoclonal antibody specifically detects CD11b in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications
Specifications
CD11b
ITGAM/3340
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified, Conjugated
ITGAM
Mouse Monoclonal CD11b Antibody (C11b/660) was immunized from recombinant fragment (around aa941-1074) of human ITGAM protein (exact sequence is proprietary)
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Alexa Fluor 350
50mM Sodium Borate with 0.05% Sodium Azide
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Mouse
IgG2a Kappa
0.1 mL
Breast Cancer, Cancer, Cell Biology, Cellular Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
Primary
Human