Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Novus Biologicals™ CD11b Recombinant Protein Antigen

Highly purified and high bioactivity. Generating reliable and reproducible results.

Applications: Antibody Competition

Manufacturer:  Novus Biologicals™ NBP234490PEP

Catalog No. NP234490PEP



Description

Description

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human CD11b

The CD11b Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.
Specifications

Specifications

3684
>80% by SDS-PAGE and Coomassie blue staining
E. coli
RUO
Antibody Competition
Unconjugated
Yes
33 kDa
100 μL
Store at -20°C. Avoid freeze-thaw cycles.
ITGAM
CD11b
Human
PBS and 1M Urea, pH 7.4.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit