Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c, APC, clone: N418, Invitrogen™ Non-distribution product as customer accommodation.

Armenian Hamster Monoclonal Antibody

Manufacturer:  Invitrogen MCD11C05

Catalog No. 50-113-7473


Add to cart

Description

Description

ITGAX is the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles.

Specifications

Specifications

CD11c
Monoclonal
APC
Itgax
Liquid
Itgax
Mouse CD11c.
Purified
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
N418
PBS with 0.1% sodium azide
Q9QXH4
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Armenian Hamster
IgG
500μL
Store at 2 - 8 °C.
Primary
16411
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit