Learn More
CD11c, Pacific Blue™, clone: BU15, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHCD11C28
Catalog No. 50-112-1334
Add to cart
Description
Description
ITGAX is the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles.
Our Pacific Blue™ dye is optimally excited by the violet laser and conjugates of this dye are strongly fluorescent even at neutral pH. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
Specifications
Specifications
CD11c
Monoclonal
Pacific Blue
ITGAX
Affinity Purified, Liquid
ITGAX
Dendritic cells of synovial fluid
Affinity chromatography
ASR, ASR/IVD
Antibody
Monoclonal
Human
Flow Cytometry
BU15
PBS with BSA, sucrose and 0.1% sodium azide
P20702
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Mouse
IgG1
0.5mL
Store reagents at 2-8°C.
Primary
3687