Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c, PE, clone: BU15, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MHCD11C04

Catalog No. 50-112-1331


Add to cart

Description

Description

ITGAX is the integrin alpha X chain protein. Integrins are heterodimeric integral membrane proteins composed of an alpha chain and a beta chain. This protein combines with the beta 2 chain (ITGB2) to form a leukocyte-specific integrin referred to as inactivated-C3b (iC3b) receptor 4 (CR4). The alpha X beta 2 complex seems to overlap the properties of the alpha M beta 2 integrin in the adherence of neutrophils and monocytes to stimulated endothelium cells, and in the phagocytosis of complement coated particles.

R-phycoerythrin (PE) is a stable and highly soluble phycobiliprotein which provides maximal absorbance and fluorescence without susceptibility to internal or external fluorescence quenching, thus providing an exceptional quantum yields and molar extinction coefficients.
Specifications

Specifications

CD11c
Monoclonal
PE
ITGAX
Liquid
ITGAX
Dendritic cells of synovial fluid
Purified
ASR, ASR/IVD
Antibody
Monoclonal
Human
Flow Cytometry
BU15
PBS with 10% sucrose, 4mg/mL BSA and 0.1% sodium azide
P20702
AI449405, Cd11c, Cr4, N418, CD11 antigen-like family member C, ITGAX, integrin aX, integrin alpha-X
Mouse
IgG1
0.5mL
Store at 2 - 8 °C.
Primary
3687
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit