Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD11c, Rabbit anti-Human,Mouse, Clone: N418, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen CD11c
Clone N418
Applications ELISA
Host Species Rabbit
Target Species Human, Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015246
View Documents
Novus Biologicals
NBP2810660.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015247
View Documents
Novus Biologicals
NBP2810660.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

CD11c Monoclonal antibody specifically detects CD11c in Human, Mouse Samples. It is validated for ELISA.
Specifications

Specifications

CD11c
ELISA
Human, Mouse
Primary
N418
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit