Learn More
CD123 Mouse, anti-Mouse, R718, Clone: 5B11, BD OptiBuild™
Mouse Monoclonal Antibody
Manufacturer:  BD Biosciences 752052
Catalog No. BDB752052
Add to cart
Description
Description
The 5B11 monoclonal antibody specifically recognizes mouse CD123, the α chain subunit (IL-3Rα) of the mouse IL-3 receptor. The IL-3Rα chain is a 60-70 kD transmembrane glycoprotein that binds IL-3 with low affinity but cannot transduce signals by itself. The mouse IL-3Rα chain can complex with either of two homologous β chain subunits (βc and βIL-3) to form high-affinity heterodimeric IL-3 receptors. The βc chain is the common β chain subunit that can complex with the α subunits of the mouse IL-3R, IL-5R and GM-CSFR to form high-affinity receptors. The βIL-3 subunit can form high affinity IL-3 receptor complexes with only the IL-3Rα subunit. The βIL-3 subunit clone is specific for mouse IL-3 and binds with low affinity. The immunogen used to generate the 5B11 hybridoma was CTLL cells transfected with the mouse IL-3R alpha chain. The 5B11 antibody does not block the binding of IL-3 protein to the high affinity IL-3 receptor heterodimer.
Specifications
Specifications
CD123
5B11
Liquid
50 μg
Refrigerate Upon Arrival
Monoclonal
Flow Cytometry
R718
Mouse
RUO
Primary
Mouse