Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD124 Mouse anti-Human, BV480, Clone: hIL4R-M57, BD OptiBuild
Mouse Monoclonal Antibody
Manufacturer: BD Biosciences 746478
Description
CD124 Monoclonal antibody specifically detects CD124 in Human samples. It is validated for Flow Cytometry.Specifications
CD124 | |
hIL4R-M57 | |
Brilliant Violet 480 | |
Mouse | |
50 μg | |
Refrigerate (2 to 8°C) | |
Monoclonal |
Flow Cytometry | |
0.2 mg/mL | |
Purified | |
IgG1, κ | |
RUO | |
Primary | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title