Learn More
CD126 Monoclonal Antibody (47.7G7.1F2), PerCP-eFluor™ 710, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen 46126942
Description
Description: This monoclonal antibody 47.7G7.1F2 reacts with human CD126, also known as IL-6 receptor alpha. It associates with the gp130 chain to form the high-affinity receptor for IL-6. Although gp130 is ubiquitously expressed, CD126 is only found on mature T cells, activated B cells, macrophages, hepatocytes, and myeloma cells. Upon binding to IL-6, two heterotrimeric complexes of receptor and ligand associate into a hexamer and are then internalized, initializing Jak/STAT signaling pathways. IL-6 induces the proliferation and survival of T cells, Th2 differentiation, and acute-phase protein production in the liver. A soluble form of CD126 lacking the transmembrane domain is also present and can be generated by cleavage at the cell surface or alternative splicing. Soluble CD126 can bind IL-6 and activate cells expressing only gp130 through trans-presentation. Applications Reported: This 47.7G7.1F2 antibody has been reported for use in flow cytometric analysis. Applications Tested: This 47.7G7.1F2 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.06 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test.
CD126 Monoclonal antibody specifically detects CD126 in Human samples. It is validated for Flow Cytometry.Specifications
CD126 | |
Monoclonal | |
5 μL/Test | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
P08887 | |
IL6R | |
Affinity chromatography | |
RUO | |
3570 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |
Flow Cytometry | |
47.7G7.1F2 | |
PerCP-eFluor 710 | |
IL6R | |
CD126; CD126 antigen; gp80; IL-6 Receptor; il-6 receptor alpha; IL-6 receptor alpha chain; IL-6 receptor subunit alpha; IL6Q; IL6R; IL-6R; IL-6R 1; IL-6R subunit alpha; IL6R1; IL-6R-1; Il6ra; IL-6RA; IL-6R-alpha; IL6RQ; ILN; Interleukin; interleukin 6 receptor; interleukin 6 receptor, alpha; interleukin-6 receptor subunit alpha; Membrane glycoprotein 80; MGC104991 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
For Research Use Only.