Learn More
CD127 (IL-7R alpha) Monoclonal Antibody (A7R34), Functional Grade, In Vivo PLATINUM™, Leinco Technologies™

Rat Monoclonal Antibody
Supplier: Leinco Technologies Inc C62231MG


Description
Specificity: Clone A7R34 recognizes an epitope on mouse CD127. Endotoxin Level: <0.01 EU/μg as determined by the LAL method. Purity: >95% monomer by analytical SEC. Preparation: Functional grade preclinical antibodies are manufactured in an animal free facility using in vitro cell culture techniques and are purified by a multi-step process including the use of protein A or G to assure extremely low levels of endotoxins, leachable protein A or aggregates. Pathogen Testing: To protect mouse colonies from infection by pathogens and to assure that experimental preclinical data is not affected by such pathogens, all of Leinco Purified Functional PLATINUM™ antibodies are tested and guaranteed to be negative for all pathogens in the IDEXX IMPACT I Mouse Profile. Storage: Functional grade preclinical antibodies may be stored sterile as received at 2-8C for up to one month. For longer term storage, aseptically aliquot in working volumes without diluting and store at -80C. Avoid Repeated Freeze Thaw Cycles.
CD127 (IL-7R alpha) Monoclonal antibody specifically detects CD127 (IL-7R alpha) in Mouse samples. It is validated for Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Inhibition Assays, Mass Spectrometry, Western BlotSpecifications
CD127 (IL-7R alpha) | |
Monoclonal | |
≥5.0 mg/mL | |
PBS with no preservative; pH 7.2-7.4 | |
P16872 | |
Rat | |
Protein A/G | |
RUO | |
16197 | |
4°C short term, -80°C long term | |
Liquid |
Flow Cytometry, Functional Assay, Immunocytochemistry, Immunohistochemistry (Frozen), Immunoprecipitation, Inhibition Assays, Mass Spectrometry, Western Blot | |
A7R34 | |
Functional Grade | |
Il7r | |
CD127; CD127 antigen; CDW127; IL 7 receptor; IL 7R a; IL 7R subunit alpha; IL 7R a; IL 7Ra; IL 7Ra; IL7 receptor; IL-7 receptor alpha chain; IL7 receptor subunit alpha; IL-7 receptor subunit alpha; IL7R; IL-7R; IL7R a; IL7R alpha; IL7R subunit alpha; IL-7R subunit alpha; IL7R a; IL7RA; IL-7RA; IL-7Ralpha; IL-7R-alpha; IL7Ra; ILRA; interleukin 7 receptor; interleukin 7 receptor alpha chain; interleukin 7 receptor isoform H5-6; Interleukin7 receptor subunit alpha; interleukin-7 receptor subunit alpha; MGC107557; sCD127; soluble IL 7 receptor; soluble IL 7R | |
IL-7Ra-IgG1 fusion protein | |
1 mg | |
Primary | |
Mouse | |
Antibody | |
IgG2a κ |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.