Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

CD127 Rat anti-Mouse, eFluor® 450, Clone: eBioSB/199 (SB/199), eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 48127380

Catalog No. 50-112-4139



Description

Description

Description: The SB/199 monoclonal antibody reacts with mouse CD127, the high affinity a subunit of the mouse IL-7 receptor. IL-7 receptor a chain is expressed by immature B cells in the bone marrow, double-negative (CD4-/CD8-), single-positive (CD4+ and CD8+), but not double-positive (CD4+/CD8+) thymocytes. In the periphery, mature T cells express low levels of CD127. Binding of SB/199 blocks the binding of A7R34, another antibody which recognizes mouse CD127. Applications Reported: This eBioSB/199 (SB/199) antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioSB/199 (SB/199) antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor® 450 is an alternative to Pacific Blue®. eFluor® 450 emits at 445 nm and is excited with the Violet laser (405 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 405 nm; Emission: 445 nm; Laser: Violet Laser. Filtration: 0.2 µm post-manufacturing filtered.

CD127 (Interleukin-7, IL-7) is a glycoprotein involved in the regulation of lymphopoiesis. The response of cells to CD127 is dependent on the presence of the interleukin 7 receptor (IL7R); the active receptor is an alpha/gamma chain heterodimer. CD127 consists of an alpha chain and a gamma chain. The gamma(c) chain, which also associates with the interleukin-2 receptor, serves primarily to activate signal transduction by the IL7R complex, while the alpha chain of IL7R determines specific signaling events through its association with cytoplasmic signaling molecules. CD127 promotes the proliferation of precursor B cells, thymocytes, T cell progenitors, and mature CD4+ and CD8+ T cells. The biological effects of IL7 are mediated by the binding of IL7 to the specific cell surface receptor. Diseases associated with CD127 dysfunction include severe combined immunodeficiency and T cell negative/B cell negative/NK positive severe combined immunodeficiency.
Specifications

Specifications

CD127
Monoclonal
0.2 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P16872
CD127, IL-7Ralpha, MGC107557
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
eBioSB/199 (SB/199)
eFluor™ 450
Il7r
Liquid
Il7r
IgG2b, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
16197
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.