Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CD13 Mouse anti-Human, BV421, Clone: L138 (also known as Leu-M7), BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 744862
Description
CD13 Monoclonal antibody specifically detects CD13 in Human samples. It is validated for Flow Cytometry.Specifications
CD13 | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
RUO | |
Human | |
Purified |
Flow Cytometry | |
L138 (also known as Leu-M7) | |
Brilliant Violet 421 | |
50 μg | |
Primary | |
Refrigerate (2 to 8°C) | |
IgG1 κ |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction