Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD132 Rat anti-Human, Biotin, Clone: TUGh4, eBioscience™ Non-distribution product as customer accommodation.

Rat Monoclonal Antibody

Manufacturer:  Invitrogen 13132980

 View more versions of this product

Catalog No. 50-117-49


Add to cart

Description

Description

Description: This TUGh4 monoclonal antibody reacts with human CD132, the common gamma chain (gamma c) shared by the IL-2, IL-4, IL-7, IL-9, and IL-15 receptors. This approximately 65 kDa receptor component is expressed on T and B lymphocytes, NK cells, monocytes, and granulocytes. CD132 is involved in intracellular signal transduction by associating with effector molecules within the cytoplasm. Mutations in the human gamma chain gene result in X-linked severe combined immunodeficiency (XSCID), a disease characterized by T cell deficiency leading to impairment of cell-mediated and humoral immunity. Applications Reported: This antibody has been reported for use in flow cytometric analysis. Applications Tested: This TUGh4 antibody has been tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at less than or equal to 0.125 µg per test. A test is defined as the amount (µg) of antibody that will stain a cell sample in a final volume of 100 µL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Filtration: 0.2 µm post-manufacturing filtered.

IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. Mutations in this gene cause X-linked severe combined immunodeficiency (XSCID), as well as X-linked combined immunodeficiency (XCID), a less severe immunodeficiency disorder.
Specifications

Specifications

CD132
Monoclonal
0.5 mg/mL
PBS with 0.1% gelatin and 0.09% sodium azide; pH 7.2
P31785
IL2RG, CIDX, IL-2RG, IMD4, P64, SCIDX, SCIDX1, CD132 antigen, IL-2 receptor subunit gamma, IL-2R subunit gamma, cytokine receptor common subunit gamma, gamma (c), gammaC, interleukin-2 receptor subunit gamma, IL2R gamma
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Human
Flow Cytometry
TUGh4
Biotin
IL2RG
Liquid
IL2RG
IgG2b, lambda
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
3561
SDS
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only.