CD133, Clone: 13A4,Millipore™ (Chemicon™)

Manufacturer: EMD Millipore MAB4310/DEL

 View more versions of this product

Catalog No. 50-175-499

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Immunogen Telencephali of ten 12-day-old NMRI mouse embryos (E12, approx. 1mg protein), which consist mostly of neuroepithelium, were homogenized in 1mL PBS, mixed with crushed nitrocellulose filter (1cm2 in 0.5mL PBS) as adjuvant, and injected i.p. into a LouXSD rat. {Weigmann et al, PNAS 1997}
    Antigen CD133
    Format Semi-purified
    Host Species Rat
    Isotype IgG1 κ
    Quantity 100μg
    Regulatory Status RUO
    Research Discipline Stem Cell Biology
    Primary or Secondary Primary
    Monoclonal or Polyclonal Monoclonal
    Concentration 1mg/mL
    Conjugate Unlabeled
    Cross Reactivity Mouse

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit